Search
Search

Col17a1| Gene Symbol: Col17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106366.1 | NP_001099836.1 | 8 - 9 | 762 | 54 | |
| XM_006231476.2 | XP_006231538.1 | 8 - 9 | 1508 | 54 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P093201 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P093204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30104 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P665039 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P093204 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P665039 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P093204 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P093204 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P665039 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P119167 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730744 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730726 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730707 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730718 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730724 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730706 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730691 | - | - |
| Rat Genome 230 2.0 Array | 1383365_at | - | - |
| Rat Genome 230 2.0 Array | 1383365_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383365_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383365_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100063474.rn.1 | TC0100007852.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383365_PM_at | - | - |
| Rat Expression Set 230 | 1383365_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21611241 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351155 | - | - |