Search
Search
| Gene Symbol: Limch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191678.1 | NP_001178607.1 | 22 - 23 | 2815 | 70 | |
| XM_006250945.2 | XP_006251007.1 | 18 - 19 | 2920 | 70 | ||
| XM_008770143.1 | XP_008768365.1 | 19 - 20 | 2989 | 70 | ||
| XM_008770144.1 | XP_008768366.1 | 19 - 20 | 2989 | 70 | ||
| XM_008770145.1 | XP_008768367.1 | 18 - 19 | 2953 | 70 | ||
| XM_008770146.1 | XP_008768368.1 | 18 - 19 | 2920 | 70 | ||
| XM_008770147.1 | XP_008768369.1 | 22 - 23 | 4388 | 70 | ||
| XM_008770148.1 | XP_008768370.1 | 19 - 20 | 2989 | 70 | ||
| XM_008770149.1 | XP_008768371.1 | 17 - 18 | 2884 | 70 | ||
| XM_008770150.1 | XP_008768372.1 | 18 - 19 | 2953 | 70 | ||
| XM_008770151.1 | XP_008768373.1 | 17 - 18 | 2884 | 70 | ||
| GenBank mRNA | FQ226534.1 | - | 11 - 12 | 1404 | 70 | |
Set Membership: |
|