Search
Search
| Gene Symbol: Cldn19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008514.1 | NP_001008514.1 | 1 - 2 | 311 | 76 | |
| XM_006238756.2 | XP_006238818.1 | 1 - 2 | 783 | 76 | ||
| XM_008763997.1 | XP_008762219.1 | 2 - 3 | 768 | 76 | ||
| GenBank mRNA | AF497645.1 | - | 1 - 2 | 227 | 76 | |
| BC079172.1 | - | 1 - 2 | 311 | 7129992 | 76 | |
Set Membership: |
|