Search
Search
| Gene Symbol: Lao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106682.1 | NP_001100152.1 | 3 - 4 | 374 | 99 | |
| GenBank mRNA | BC100634.1 | - | 3 - 4 | 374 | 99 | |
| BC168679.1 | - | 3 - 4 | 374 | 5621187 | 99 | |
Set Membership: |
|