Search
Search

Tmem221| Gene Symbol: Tmem221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728393.4 | XP_002728439.2 | 1 - 2 | 1101 | 70 | |
| XM_006252969.2 | XP_006253031.1 | 1 - 2 | 1101 | 70 | ||
Set Membership: |
|