Search
Search
| Gene Symbol: Sys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001062200.5 | XP_001062200.2 | 2 - 3 | 482 | 123 | |
| XM_008762556.1 | XP_008760778.1 | 1 - 2 | 386 | 123 | ||
| XM_008762557.1 | XP_008760779.1 | 1 - 2 | 395 | 123 | ||
Set Membership: |
|