Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014225.1 | NP_001014247.1 | 5 - 6 | 834 | 71 | |
XM_006245832.2 | XP_006245894.1 | 5 - 6 | 597 | 71 | ||
XM_008767512.1 | XP_008765734.1 | 5 - 6 | 530 | 71 | ||
XM_008767513.1 | XP_008765735.1 | 5 - 6 | 892 | 71 | ||
XM_008767516.1 | XP_008765738.1 | 4 - 5 | 436 | 71 | ||
XM_008767517.1 | XP_008765739.1 | 3 - 4 | 299 | 71 | ||
XM_008767518.1 | XP_008765740.1 | 3 - 4 | 356 | 71 | ||
GenBank mRNA | BC079022.1 | - | 5 - 6 | 834 | 7113868 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |