Search
Search
| Gene Symbol: Rpusd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108641.1 | NP_001102111.1 | 6 - 7 | 869 | 94 | |
| XM_006237044.2 | XP_006237106.1 | 7 - 8 | 734 | 94 | ||
| XM_006237045.2 | XP_006237107.1 | 7 - 8 | 909 | 94 | ||
| XM_006237046.2 | XP_006237108.1 | 7 - 8 | 734 | 94 | ||
Set Membership: |
|