Search
Search
| Gene Symbol: Lmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236972.2 | XP_006237034.1 | 2 - 3 | 526 | 93 | |
| XM_006236973.2 | XP_006237035.1 | 2 - 3 | 413 | 93 | ||
| XM_575617.7 | XP_575617.4 | 1 - 2 | 518 | 93 | ||
Set Membership: |
|