Search
Search
| Gene Symbol: Phf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212538.1 | NP_997703.1 | 7 - 8 | 897 | 59 | |
| XM_006256117.2 | XP_006256179.1 | 7 - 8 | 1151 | 59 | ||
| XM_008772723.1 | XP_008770945.1 | 7 - 8 | 1152 | 59 | ||
| GenBank mRNA | BC083843.1 | - | 7 - 8 | 838 | 7114670 | 59 |
Set Membership: |
|