Search
Search
| Gene Symbol: Tead3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098216.1 | NP_001091686.1 | 9 - 10 | 973 | 67 | |
| GenBank mRNA | BC085938.1 | - | 9 - 10 | 926 | 67 | |
| BC099077.1 | - | 9 - 10 | 973 | 67 | ||
Set Membership: |
|