Search
Search
| Gene Symbol: Syne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227853.2 | XP_006227915.1 | 28 - 29 | 4133 | 61 | |
| XM_008758714.1 | XP_008756936.1 | 27 - 28 | 4119 | 61 | ||
| XM_008758715.1 | XP_008756937.1 | 29 - 30 | 4186 | 61 | ||
| XM_008758716.1 | XP_008756938.1 | 29 - 30 | 4184 | 61 | ||
| GenBank mRNA | AY597251.1 | - | 15 - 16 | 2529 | 61 | |
| X95466.1 | - | 14 - 15 | 2441 | 61 | ||
Set Membership: |
|