Search
Search

Ccdc107| Gene Symbol: Ccdc107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749929.3 | XP_003749977.1 | 4 - 5 | 501 | 62 | |
| XM_006238135.2 | XP_006238197.1 | 4 - 5 | 513 | 62 | ||
| XM_006238136.2 | XP_006238198.1 | 4 - 5 | 509 | 62 | ||
| XM_006238137.2 | XP_006238199.1 | 4 - 5 | 505 | 62 | ||
| GenBank mRNA | FQ229548.1 | - | 5 - 6 | 510 | 62 | |
Set Membership: |
|