Search
Search
| Gene Symbol: Srpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106575.2 | NP_001100045.1 | 3 - 4 | 492 | 57 | |
| XM_006235903.2 | XP_006235965.1 | 3 - 4 | 3488 | 57 | ||
| XM_008762635.1 | XP_008760857.1 | 3 - 4 | 2910 | 57 | ||
| XM_008762636.1 | XP_008760858.1 | 3 - 4 | 3488 | 57 | ||
| XM_008762637.1 | XP_008760859.1 | 3 - 4 | 3487 | 57 | ||
| GenBank mRNA | BC161879.1 | - | 3 - 4 | 492 | 7125533 | 57 |
| FQ234485.1 | - | 3 - 4 | 505 | 57 | ||
Set Membership: |
|