Search
Search
| Gene Symbol: Gsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107845.1 | NP_001101315.1 | 1 - 2 | 163 | 65 | |
| XM_006235935.2 | XP_006235997.1 | 1 - 2 | 217 | 65 | ||
| XM_006235936.2 | XP_006235998.1 | 1 - 2 | 219 | 65 | ||
| XM_008762648.1 | XP_008760870.1 | 1 - 2 | 219 | 65 | ||
| XM_008762649.1 | XP_008760871.1 | 1 - 2 | 218 | 65 | ||
Set Membership: |
|