Search
Search
| Gene Symbol: Usp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001516.2 | NP_001001516.1 | 7 - 8 | 1104 | 56 | |
| XM_006243793.2 | XP_006243855.1 | 8 - 9 | 1416 | 56 | ||
| XM_006243794.2 | XP_006243856.1 | 8 - 9 | 1413 | 56 | ||
| XM_006243795.2 | XP_006243857.1 | 8 - 9 | 1416 | 56 | ||
| XM_006243796.2 | XP_006243858.1 | 8 - 9 | 1392 | 56 | ||
| XM_006243797.2 | XP_006243859.1 | 8 - 9 | 1415 | 56 | ||
| XM_006243798.2 | XP_006243860.1 | 8 - 9 | 1412 | 56 | ||
| XM_006243799.2 | XP_006243861.1 | 8 - 9 | 1415 | 56 | ||
| GenBank mRNA | AY605065.1 | - | 7 - 8 | 1104 | 56 | |
| BC168243.1 | - | 7 - 8 | 1424 | 56 | ||
Set Membership: |
|