Search
Search
| Gene Symbol: Snap91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031728.1 | NP_113916.1 | 7 - 8 | 712 | 149 | |
| XM_006243491.2 | XP_006243553.1 | 7 - 8 | 849 | 149 | ||
| XM_006243492.2 | XP_006243554.1 | 7 - 8 | 910 | 149 | ||
| XM_006243493.2 | XP_006243555.1 | 7 - 8 | 849 | 149 | ||
| XM_006243494.2 | XP_006243556.1 | 7 - 8 | 849 | 149 | ||
| XM_006243495.2 | XP_006243557.1 | 7 - 8 | 849 | 149 | ||
| XM_006243496.2 | XP_006243558.1 | 7 - 8 | 850 | 149 | ||
| XM_006243497.2 | XP_006243559.1 | 7 - 8 | 849 | 149 | ||
| XM_008766455.1 | XP_008764677.1 | 7 - 8 | 805 | 149 | ||
| XM_008766456.1 | XP_008764678.1 | 7 - 8 | 849 | 149 | ||
| GenBank mRNA | X68877.1 | - | 7 - 8 | 712 | 149 | |
| X68878.1 | - | 7 - 8 | 712 | 149 | ||
Set Membership: |
|