Search
Search

LOC690507| Gene Symbol: LOC690507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235368.2 | XP_006235430.2 | 6 - 7 | 1298 | 68 | |
| XM_008762403.1 | XP_008760625.1 | 7 - 8 | 2885 | 68 | ||
Set Membership: |
|