Search
Search

Cdc42bpg| Gene Symbol: Cdc42bpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130013.1 | NP_001123485.1 | 5 - 6 | 837 | 78 | |
| XM_006230751.2 | XP_006230813.1 | 5 - 6 | 776 | 78 | ||
| XM_006230752.2 | XP_006230814.1 | 5 - 6 | 706 | 78 | ||
| XM_008760108.1 | XP_008758330.1 | 5 - 6 | 487 | 78 | ||
Set Membership: |
|