Search
Search
| Gene Symbol: Fscn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100806.1 | NP_001094276.1 | 3 - 4 | 1366 | 58 | |
| GenBank mRNA | AY268934.1 | - | 200 | 58 | ||
| BC168678.1 | - | 3 - 4 | 1366 | 5598674 | 58 | |
Set Membership: |
|