Search
Search
| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053304.1 | NP_445756.1 | 1 - 2 | 189 | 115 | |
| GenBank mRNA | BC133728.1 | - | 1 - 2 | 173 | 7125649 | 115 |
| CB693258.1 | - | 1 - 2 | 197 | 115 | ||
| Z78279.1 | - | 1 - 2 | 78 | 115 | ||
Set Membership: |
|