Search
Search

Ccdc64b| Gene Symbol: Ccdc64b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106982.1 | NP_001100452.1 | 5 - 6 | 845 | 69 | |
| GenBank mRNA | BC161878.1 | - | 5 - 6 | 793 | 7123846 | 69 |
Set Membership: |
|