Search
Search

Ccdc137| Gene Symbol: Ccdc137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143896.1 | NP_001137368.1 | 1 - 2 | 165 | 154 | |
| XM_008768483.1 | XP_008766705.1 | 1 - 2 | 205 | 154 | ||
| GenBank mRNA | BC158793.1 | - | 1 - 2 | 132 | 154 | |
Set Membership: |
|