Search
Search

LOC497940| Gene Symbol: LOC497940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017476.1 | NP_001017476.1 | 2 - 3 | 678 | 74 | |
| XM_006246777.2 | XP_006246839.1 | 2 - 3 | 825 | 74 | ||
| XM_006246778.2 | XP_006246840.1 | 2 - 3 | 829 | 74 | ||
| XM_006246781.2 | XP_006246843.1 | 2 - 3 | 826 | 74 | ||
| XM_006246782.2 | XP_006246844.1 | 2 - 3 | 822 | 74 | ||
| GenBank mRNA | BC079326.1 | - | 2 - 3 | 678 | 7135484 | 74 |
Set Membership: |
|