Search
Search
| Gene Symbol: Nle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127534.1 | NP_001121006.1 | 10 - 11 | 1226 | 115 | |
| XM_006246994.2 | XP_006247056.1 | 11 - 12 | 2066 | 115 | ||
| XM_006246995.2 | XP_006247057.1 | 11 - 12 | 1936 | 115 | ||
| GenBank mRNA | BC166567.1 | - | 10 - 11 | 1226 | 7317666 | 115 |
Set Membership: |
|