Search
Search

Arhgap27| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198759.1 | NP_942054.1 | 9 - 10 | 1784 | 82 | |
| XM_006247523.2 | XP_006247585.1 | 11 - 12 | 2907 | 82 | ||
| XM_006247525.2 | XP_006247587.1 | 9 - 10 | 2280 | 82 | ||
| XM_008768290.1 | XP_008766512.1 | 11 - 12 | 3203 | 82 | ||
| XM_008768291.1 | XP_008766513.1 | 9 - 10 | 1275 | 82 | ||
| GenBank mRNA | AY394725.1 | - | 9 - 10 | 1784 | 82 | |
Set Membership: |
|