Search
Search
| Gene Symbol: Lclat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764536.1 | XP_008762758.1 | 2 - 3 | 7770 | 96 | |
| XM_008764537.1 | XP_008762759.1 | 2 - 3 | 320 | 96 | ||
| XM_008764538.1 | XP_008762760.1 | 2 - 3 | 310 | 96 | ||
| XM_008764539.1 | XP_008762761.1 | 2 - 3 | 7770 | 96 | ||
Set Membership: |
|