Search
Search

Atxn7l4| Gene Symbol: Atxn7l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240009.2 | XP_006240071.1 | 5 - 6 | 900 | 94 | |
| XM_006240013.2 | XP_006240075.1 | 4 - 5 | 1172 | 94 | ||
| XM_006240014.2 | XP_006240076.1 | 4 - 5 | 1380 | 94 | ||
| XM_006240015.2 | XP_006240077.1 | 5 - 6 | 900 | 94 | ||
| XM_006240016.2 | XP_006240078.1 | 5 - 6 | 900 | 94 | ||
| XM_006240017.2 | XP_006240079.1 | 3 - 4 | 553 | 94 | ||
| XM_006240018.2 | XP_006240080.1 | 2 - 3 | 437 | 94 | ||
| XM_008764632.1 | XP_008762854.1 | 5 - 6 | 900 | 94 | ||
| XM_008764633.1 | XP_008762855.1 | 4 - 5 | 2255 | 94 | ||
| XM_008764634.1 | XP_008762856.1 | 5 - 6 | 900 | 94 | ||
| XM_008764635.1 | XP_008762857.1 | 2 - 3 | 1076 | 94 | ||
Set Membership: |
|