Search
Search
| Gene Symbol: Kdm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108829.1 | NP_001102299.1 | 8 - 9 | 3245 | 86 | |
| GenBank mRNA | EF532390.1 | - | 3 - 4 | 919 | 86 | |
| EF532391.1 | - | 3 - 4 | 906 | 86 | ||
| EF532392.1 | - | 3 - 4 | 919 | 86 | ||
| EF532393.1 | - | 3 - 4 | 906 | 86 | ||
Set Membership: |
|