Search
Search
| Gene Symbol: Ric1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231255.2 | XP_006231317.1 | 22 - 23 | 3631 | 92 | |
| XM_006231256.2 | XP_006231318.1 | 22 - 23 | 3629 | 92 | ||
| XM_219778.8 | XP_219778.5 | 22 - 23 | 3619 | 92 | ||
Set Membership: |
|