Search
Search

RGD1310371| Gene Symbol: RGD1310371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100581.1 | NP_001094051.1 | 1 - 2 | 78 | 74 | |
| GenBank mRNA | BC158775.1 | - | 1 - 2 | 78 | 7132188 | 74 |
Set Membership: |
|