Search
Search
| Gene Symbol: Rasal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108335.1 | NP_001101805.1 | 16 - 17 | 1899 | 56 | |
| XM_006249395.2 | XP_006249457.1 | 15 - 16 | 3096 | 56 | ||
| XM_006249396.2 | XP_006249458.1 | 15 - 16 | 3100 | 56 | ||
Set Membership: |
|