Search
Search

RGD1566099| Gene Symbol: RGD1566099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108347.1 | NP_001101817.1 | 2 - 3 | 364 | 161 | |
| GenBank mRNA | BC166874.1 | - | 2 - 3 | 361 | 6890641 | 161 |
| BC166876.1 | - | 2 - 3 | 361 | 6919569 | 161 | |
| BC166924.1 | - | 2 - 3 | 361 | 6919449 | 161 | |
Set Membership: |
|