Search
Search
| Gene Symbol: Ptgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017232.3 | NP_058928.3 | 3 - 4 | 312 | 112 | |
| GenBank mRNA | AF233596.1 | - | 436 | 112 | ||
| AY157736.1 | - | 3 - 4 | 312 | 112 | ||
| L20085.1 | - | 3 - 4 | 312 | 112 | ||
| L25925.1 | - | 3 - 4 | 318 | 112 | ||
| S67722.1 | - | 436 | 112 | |||
| U03389.1 | - | 436 | 112 | |||
| U04300.1 | - | 3 - 4 | 312 | 112 | ||
Set Membership: |
|