Search
Search
| Gene Symbol: Commd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033692.1 | NP_001028864.1 | 2 - 3 | 183 | 80 | |
| XM_006234637.2 | XP_006234699.1 | 2 - 3 | 218 | 80 | ||
| GenBank mRNA | BC101873.1 | - | 2 - 3 | 183 | 5621281 | 80 |
| EU000469.1 | - | 2 - 3 | 179 | 80 | ||
| FQ213255.1 | - | 3 - 4 | 267 | 80 | ||
Set Membership: |
|