Search
Search
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011906.1 | NP_001011906.1 | 2 - 3 | 790 | 104 | |
| XM_006250031.2 | XP_006250093.1 | 2 - 3 | 846 | 104 | ||
| XM_006250032.2 | XP_006250094.1 | 2 - 3 | 854 | 104 | ||
| XM_006250033.2 | XP_006250095.1 | 2 - 3 | 853 | 104 | ||
| XM_006250035.2 | XP_006250097.1 | 2 - 3 | 847 | 104 | ||
| XM_006250036.2 | XP_006250098.1 | 2 - 3 | 847 | 104 | ||
| GenBank mRNA | BC086598.1 | - | 2 - 3 | 790 | 7313105 | 104 |
Set Membership: |
|