Search
Search

RGD1563746| Gene Symbol: RGD1563746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247481.2 | XP_006247543.1 | 5 - 6 | 2526 | 109 | |
| XM_220997.8 | XP_220997.5 | 5 - 6 | 2586 | 109 | ||
Set Membership: |
|