Search
Search
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106587.1 | NP_001100057.1 | 7 - 8 | 1364 | 60 | |
| XM_006236186.2 | XP_006236248.1 | 8 - 9 | 1138 | 60 | ||
| GenBank mRNA | AY724521.1 | - | 7 - 8 | 1138 | 60 | |
Set Membership: |
|