Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500969_m1
          See other LOC500034 GE Assays ›
          Gene Symbol:
          LOC500034
          Gene Name:
          similar to CG3570-PA
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 40064131 - 40136074 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01500969_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC500034
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109221.2 NP_001102691.1 4 - 5 578 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002476.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA methylation
          rRNA (adenine) methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P464684 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81952 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P464684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P464684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P464684 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18902 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100926 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861120 - -
          Affymetrix Rat Gene 1.0 ST Array 10861121 - -
          Affymetrix Rat Gene 1.0 ST Array 10861122 - -
          Affymetrix Rat Gene 1.0 ST Array 10861118 - -
          Affymetrix Rat Gene 1.0 ST Array 10861123 - -
          Affymetrix Rat Gene 1.0 ST Array 10861119 - -
          Affymetrix Rat Gene 1.1 ST Array 10861117 - -
          Rat Genome 230 2.0 Array 1389558_PM_at - -
          Rat Genome 230 2.0 Array 1389558_at - -
          Rat Genome 230 2.0 Array 1390394_at - -
          Rat Genome 230 2.0 Array 1390394_PM_at - -
          Affymetrix HT Rat Focus Array 1390394_at - -
          Affymetrix HT Rat Focus Array 1390394_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400017621.rn.1 TC0400002476.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017623.rn.1 TC0400002476.rn.1 RTA TC
          Rat Expression Set 230 1390394_at - -
          Rat Expression Set 230 1389558_at - -
          Rat Expression Set 230 1390394_PM_at - -
          Rat Expression Set 230 1389558_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10861117 - -
          Rat Genome U34 Set rc_AI012073_at - -
          Rat Genome U34 Set rc_AI233529_i_at - -
          Rat Genome U34 Set rc_AI233529_f_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline