Search
Search
| Gene Symbol: Tcrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC091277.1 | - | 859 | 65 | ||
| BC091428.1 | - | 945 | 7367078 | 65 | ||
| BC099166.1 | - | 987 | 7382105 | 65 | ||
| BC105831.1 | - | 857 | 65 | |||
| BC107948.1 | - | 802 | 65 | |||
| BC158832.1 | - | 855 | 7380746 | 65 | ||
| X14319.1 | - | 681 | 65 | |||
Set Membership: |
|