Search
Search
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108088.1 | NP_001101558.1 | 5 - 6 | 622 | 144 | |
| XM_006241286.2 | XP_006241348.1 | 4 - 5 | 985 | 144 | ||
| GenBank mRNA | BC161903.1 | - | 4 - 5 | 828 | 8367209 | 144 |
Set Membership: |
|