Search
Search

RGD1306001| Gene Symbol: RGD1306001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130696.1 | NP_001124168.1 | 2 - 3 | 228 | 99 | |
| XM_006242163.1 | XP_006242225.1 | 2 - 3 | 804 | 99 | ||
| XM_008765765.1 | XP_008763987.1 | 2 - 3 | 392 | 99 | ||
Set Membership: |
|