Search
Search
| Gene Symbol: Cyp2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138515.2 | NP_612524.1 | 2 - 3 | 375 | 77 | |
| GenBank mRNA | AB008425.1 | - | 1 - 2 | 248 | 77 | |
| BC093609.1 | - | 390 | 7319137 | 77 | ||
| FQ211703.1 | - | 391 | 77 | |||
| U48219.1 | - | 2 - 3 | 375 | 77 | ||
| U48220.1 | - | 373 | 77 | |||
Set Membership: |
|