Search
Search

LOC102554586| Gene Symbol: LOC102554586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759935.1 | XP_008758157.1 | 2 - 3 | 203 | 116 | |
| GenBank mRNA | BG372695.1 | - | 1 - 2 | 116 | ||
Set Membership: |
|