Search
Search
| Gene Symbol: Dgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012345.1 | NP_001012345.1 | 2 - 3 | 423 | 74 | |
| XM_008759689.1 | XP_008757911.1 | 2 - 3 | 174 | 74 | ||
| GenBank mRNA | BC089846.1 | - | 2 - 3 | 423 | 7372077 | 74 |
Set Membership: |
|