Search
Search

RGD1563217| Gene Symbol: RGD1563217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109155.1 | NP_001102625.1 | 2 - 3 | 231 | 90 | |
| XM_006230320.2 | XP_006230382.1 | 2 - 3 | 409 | 90 | ||
| XM_006230321.2 | XP_006230383.1 | 2 - 3 | 188 | 90 | ||
Set Membership: |
|