Search
Search
| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177688.1 | NP_001171159.1 | 5 - 6 | 724 | 68 | |
| XM_006234347.2 | XP_006234409.1 | 5 - 6 | 796 | 68 | ||
| GenBank mRNA | BC160919.1 | - | 5 - 6 | 724 | 7458373 | 68 |
Set Membership: |
|