Search
Search
| Gene Symbol: Rpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033999.2 | NP_001029171.1 | 7 - 8 | 1016 | 60 | |
| XM_006234771.2 | XP_006234833.1 | 7 - 8 | 1073 | 60 | ||
| XM_006234772.2 | XP_006234834.1 | 6 - 7 | 962 | 60 | ||
| XM_006234773.2 | XP_006234835.1 | 6 - 7 | 941 | 60 | ||
Set Membership: |
|