Search
Search
| Gene Symbol: Pcnx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170347.1 | NP_001163818.1 | 18 - 19 | 3788 | 59 | |
| XM_006240299.2 | XP_006240361.1 | 18 - 19 | 3799 | 59 | ||
| XM_006240300.2 | XP_006240362.1 | 18 - 19 | 3796 | 59 | ||
| XM_006240301.1 | XP_006240363.1 | 19 - 20 | 3835 | 59 | ||
| XM_006240302.2 | XP_006240364.1 | 16 - 17 | 3481 | 59 | ||
| XM_008764788.1 | XP_008763010.1 | 17 - 18 | 3709 | 59 | ||
| XM_008764789.1 | XP_008763011.1 | 18 - 19 | 5195 | 59 | ||
Set Membership: |
|