Search
Search
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108695.1 | NP_001102165.1 | 2 - 3 | 386 | 91 | |
| GenBank mRNA | BC159427.1 | - | 2 - 3 | 284 | 9088591 | 91 |
| FQ229895.1 | - | 2 - 3 | 272 | 91 | ||
Set Membership: |
|